1H,1H,2H,2H-Perfluorooctyltrichlorosilane ≥97.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
Tridecafluoro-1,1,2,2-tetrahydrooctyl-1-trichlorosilane
T2577-5G
T2577-25G
TCIAT2577-5GEA
72
EUR
TCIAT2577-5G
TCIAT2577-25G
1H,1H,2H,2H-Perfluorooctyltrichlorosilane ≥97.0% (by GC)
1H,1H,2H,2H-Perfluorooctyltrichlorosilane
Formula:
C₈H₄Cl₃F₁₃Si Peso molecolare: 481,55 g/mol Punto di ebollizione: 85°C Flash Pt: 54°C |
MDL Number:
MFCD00042363 CAS Number: 78560-45-9 EINECS: 278-947-6 UN: 2986 ADR: 8,II |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...