2,2',4,4'-Tetramethylbenzophenone ≥96.0% (by GC)
Fornitore: TCI
Formula:
C₁₇H₁₈O Punto di ebollizione: 188°C |
MDL Number:
MFCD00060094 CAS Number: 3478-88-4 |
|
Risultati del test di specificazione
Appearance | Colorless to Yellow to Green clear liquid |
Purity (GC) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...