4-(Boc-amino)piperidine ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
4-N-(tert-Butoxycarbonyl)aminopiperidine, tert-Butyl piperidin-4-ylcarbamate, tert-butyl N-(piperidin-4-yl)carbamate, 4-Boc-Aminopiperidine, 4-N-BOC-Aminopiperidine
B2319-1G
B2319-5G
TCIAB2319-1GEA
29
EUR
TCIAB2319-1G
TCIAB2319-5G
4-(Boc-amino)piperidine ≥98.0% (by GC, titration analysis)
4-(Boc-amino)piperidine
Formula:
C₁₀H₂₀N₂O₂ Peso molecolare: 200,28 g/mol |
MDL Number:
MFCD00798171 CAS Number: 73874-95-0 |
Risultati del test di specificazione
Appearance | White to Light yellow to Light red powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 158.0 to 162.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...