Stampa…

(S)-3-(Boc-amino)piperidine ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: (S)-3-Boc-aminopiperidine, (S)-tert-Butyl piperidin-3-ylcarbamate, tert-butyl N-[(3S)-piperidin-3-yl]carbamate

B3660-1G B3660-5G
TCIAB3660-1GEA 23 EUR
TCIAB3660-1G TCIAB3660-5G
(S)-3-(Boc-amino)piperidine ≥98.0% (by GC)
(S)-3-(Boc-amino)piperidine
Formula: C₁₀H₂₀N₂O₂
Peso molecolare: 200,28 g/mol
MDL Number: MFCD03093383
CAS Number: 216854-23-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7f107fc4db157eb60e29bc2c43a55e17e21fd020&vid=7f107fc4db157eb60e29bc2c43a55e17e21fd020&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 118.0 to 125.0 deg-C
Specific rotation [a]20/D -12.0 to -16.0 deg (C=1, EtOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR