(R)-3-(Boc-amino)piperidine ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
tert-Butyl N-[(3R)-piperidin-3-yl]carbamate, (R)-3-Boc-aminopiperidine, (R)-tert-Butyl piperidin-3-ylcarbamate
B3591-1G
B3591-5G
TCIAB3591-1GEA
39
EUR
TCIAB3591-1G
TCIAB3591-5G
(R)-3-(Boc-amino)piperidine ≥98.0% (by GC, titration analysis)
(R)-3-(Boc-amino)piperidine
Formula:
C₁₀H₂₀N₂O₂ Peso molecolare: 200,28 g/mol |
MDL Number:
MFCD03093382 CAS Number: 309956-78-3 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 121.0 to 125.0 deg-C |
Specific rotation [a]20/D | +13.0 to +16.0 deg (C=1, EtOH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...