Stampa…

7-(2-Hydroxyethyl)theophylline ≥98.0% (by HPLC, total nitrogen)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 7-(2-Hydroxyethyl)-1,3-dimethyl-3,7-dihydro-1H-purine-2,6-dione, Etofylline

H0402-25G H0402-500G
TCIAH0402-25GEA 48 EUR
TCIAH0402-25G TCIAH0402-500G
7-(2-Hydroxyethyl)theophylline ≥98.0% (by HPLC, total nitrogen)
7-(2-Hydroxyethyl)theophylline
Formula: C₉H₁₂N₄O₃
Peso molecolare: 224,216 g/mol
MDL Number: MFCD00055055
CAS Number: 519-37-9
EINECS: 208-269-8
Merck Index: 14,03878
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=97d0d1875552c1b94b944c38da3cd5aa2f155c9b&vid=97d0d1875552c1b94b944c38da3cd5aa2f155c9b&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 162.0 to 166.0 deg-C
Solubility in hot Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR