Stampa…
D-(-)-Luciferin sodium salt monohydrate
Fornitore: Biotium
10102-2 10102 10102-1
10102-2EA 1190 EUR
10102-2 10102. 10102-1
D-(-)-Luciferin sodium salt monohydrate
D-(-)-Luciferin sodium salt monohydrate
Formula: C₁₁H₇N₂NaO₃S₂·H₂O
Peso molecolare: 320,32 g/mol
MDL Number: MFCD11865368
CAS Number: 103404-75-7