Diosmin ≥85.0% (by HPLC)
Fornitore: TCI
D3908-25G
D3908-5G
TCIAD3908-25GEA
101
EUR
TCIAD3908-25G
TCIAD3908-5G
Diosmin ≥85.0% (by HPLC)
Diosmin
Formula:
C₂₈H₃₂O₁₅ Punto di fusione: 280°C |
MDL Number:
MFCD00009772 CAS Number: 520-27-4 EINECS: 208-289-7 Merck Index: 14,03297 |
|
Risultati del test di specificazione
Appearance | White to Gray to Brown powder to crystal |
Purity (HPLC) | min. 85.0 area% |
Specific rotation [a]20/D | -116 ~ -126 deg (C=0.5, 0.1mol/L NaOH sol.) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...