Stampa…

Carbamazepine ≥97.0% (by total nitrogen basis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 5H-Dibenzo[b,f]azepine-5-carboxamide

C1095-10G C1095-25G
TCIAC1095-10GEA 51 EUR
TCIAC1095-10G TCIAC1095-25G
Carbamazepine ≥97.0% (by total nitrogen basis)
Carbamazepine
Formula: C₁₅H₁₂N₂O
Peso molecolare: 236,27 g/mol
MDL Number: MFCD00005073
CAS Number: 298-46-4
EINECS: 206-062-7
Merck Index: 14,01781
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8eb2db8a8884385766f478f1e0a39bb7f0dd8c2a&vid=8eb2db8a8884385766f478f1e0a39bb7f0dd8c2a&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 97.0 %
Melting point 188.0 to 193.0 deg-C
Solubility in Methanol within very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR