Stampa…

2-Methyl-4'-(methylthio)-2-morpholinopropiophenone ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 2-Methyl-1-(4-(methylthio)phenyl)-2-morpholinopropan-1-one, 2-Methyl-1-[4-(methylthio)phenyl]-2-morpholino-1-propanone

M2028-25G M2028-500G
TCIAM2028-25GEA 24 EUR
TCIAM2028-25G TCIAM2028-500G
2-Methyl-4'-(methylthio)-2-morpholinopropiophenone ≥98.0% (by GC, titration analysis)
2-Methyl-4'-(methylthio)-2-morpholinopropiophenone
Formula: C₁₅H₂₁NO₂S
Peso molecolare: 279,3977 g/mol
Punto di ebollizione: 210°C
MDL Number: MFCD00083014
CAS Number: 71868-10-5
EINECS: 400-600-6
UN: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0ff6430dfcece8e39f5626ab13058fad80c49322&vid=0ff6430dfcece8e39f5626ab13058fad80c49322&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 73.0 to 77.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR