Stampa…

Tin(II) phthalocyanine

Fornitore: TCI EUROPE
Pericolo

Synonyms: Tin(ii)phthalocyanine

P1024-1G P1024-5G
TCIAP1024-1GEA 62 EUR
TCIAP1024-1G TCIAP1024-5G
Tin(II) phthalocyanine
Tin(II) phthalocyanine
Formula: C₃₂H₁₆N₈Sn
Peso molecolare: 631,233 g/mol
MDL Number: MFCD00058867
CAS Number: 15304-57-1
UN: 3146
ADR: 6.1,III

Visualizza i codici

Risultati del test di specificazione

Appearance Gray to Dark purple to Black powder to crystal
Ignition residue (as SnO2) 22.7 to 24.3 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR