Stampa…

3-Aminopropionitrile fumarate ≥98.0% (by titrimetric analysis)

Fornitore: TCI
Pericolo

A0796-25G A0796-500G
TCIAA0796-25GEA 50 EUR
TCIAA0796-25G TCIAA0796-500G
3-Aminopropionitrile fumarate ≥98.0% (by titrimetric analysis)
3-Aminopropionitrile fumarate
Formula: C₁₀H₁₆N₄O₄
MDL Number: MFCD00013142
CAS Number: 2079-89-2
EINECS: 218-208-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bd0f74426efac3f245ba85d31012d6ab5e66a628&vid=29bba65eb8740dbb146ae705a2fb0f777cbaa24e&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR