Stampa…

Phenoxyacetic anhydride ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-Phenoxyacetic anhydride

P0111-25G P0111-250G
TCIAP0111-25GEA 119 EUR
TCIAP0111-25G TCIAP0111-250G
Phenoxyacetic anhydride ≥98.0% (by titrimetric analysis)
Phenoxyacetic anhydride
Formula: C₁₆H₁₄O₅
Peso molecolare: 286,2794 g/mol
MDL Number: MFCD00059681
CAS Number: 14316-61-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c6c46965d8a1797c37596bfdb5d34cbccd456dc1&vid=c6c46965d8a1797c37596bfdb5d34cbccd456dc1&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Anilin Method) min. 98.0 %
Melting point 71.0 to 74.0 deg-C
Solubility in Toluene almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR