N-(2-Hydroxyethyl)iminodiacetic acid ≥98.0% (by titrimetric analysis)
Fornitore: TCI
|
Attenzione
|
Synonyms:
2,2'-((2-Hydroxyethyl)azanediyl)diacetic acid
Formula:
C₆H₁₁NO₅ Peso molecolare: 177.155 g/mol |
MDL Number:
MFCD00004293 CAS Number: 93-62-9 EINECS: 202-263-9 |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (Chelometric Titration) | min. 98.0 % |
Solubility in hot Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...