Stampa…

N-Methyliminodiacetic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

M2090-5G M2090-25G
TCIAM2090-5GEA 16 EUR
TCIAM2090-5G TCIAM2090-25G
N-Methyliminodiacetic acid ≥98.0% (by GC, titration analysis)
N-Methyliminodiacetic acid
Formula: C₅H₉NO₄
Punto di fusione: 219°C
MDL Number: MFCD00004284
CAS Number: 4408-64-4
EINECS: 224-557-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5cc892a2e05448b8fbf39ed3cd1daa33e5fb5a2d&vid=5cc892a2e05448b8fbf39ed3cd1daa33e5fb5a2d&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR