Stampa…

N-Methyliminodiacetic acid 99%

Fornitore: Thermo Fisher Scientific
Attenzione

127010050 127010250
ACRO127010050EA 39.1 EUR
ACRO127010050 ACRO127010250
N-Methyliminodiacetic acid 99%
N-Methyliminodiacetic acid
Formula: C₅H₉NO₄
Punto di fusione: 218…221 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00004284
CAS Number: 4408-64-4
EINECS: 224-557-6

Visualizza i codici

Risultati del test di specificazione

Appearance White to cream Powder
Infrared spectrum Conforms
Titration with NaOH ≥98.5 %
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR