Stampa…

N-Phenylglycine ≥97.0% (by titrimetric analysis)

Fornitore: TCI

P0180-25G P0180-100G
TCIAP0180-25GEA 48 EUR
TCIAP0180-25G TCIAP0180-100G
N-Phenylglycine ≥97.0% (by titrimetric analysis)
N-Phenylglycine
Formula: C₈H₉NO₂
MDL Number: MFCD00014009
CAS Number: 103-01-5
EINECS: 203-070-2
Merck Index: 14,07292
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=526621d9c3df57dc0b2a597af4f06e3e214d7dac&vid=526621d9c3df57dc0b2a597af4f06e3e214d7dac&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Green to Brown powder to crystal
Purity (Neutralization titration) min. 97.0 %
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR