Stampa…

L(+)-Diisopropyl tartrate ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: Diisopropyl (2R,3R)-2,3-dihydroxysuccinate, L-(+)-Tartaric acid diisopropyl ester

T0621-25G T0621-500G
TCIAT0621-25GEA 19 EUR
TCIAT0621-25G TCIAT0621-500G
L(+)-Diisopropyl tartrate ≥98.0% (by GC)
L(+)-Diisopropyl tartrate
Formula: C₁₀H₁₈O₆
Peso molecolare: 234,25 g/mol
Punto di ebollizione: 275°C
Flash Pt: 110°C
MDL Number: MFCD00064450
CAS Number: 2217-15-4
EINECS: 218-709-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7a6d7441f49d81a0884f8b39c2f25a48bc8397c7&vid=7a6d7441f49d81a0884f8b39c2f25a48bc8397c7&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Optical purity (GC) min. 99.0 ee%
Specific rotation [a]20/D +16.3 to +16.6 deg (neat)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR