N-Boc-glycine (boc-gly-OH) ≥98.0% (by titrimetric analysis)
Fornitore: TCI
Synonyms:
Boc-Gly-OH
B1185-25G
B1185-5G
TCIAB1185-25GEA
42
EUR
TCIAB1185-25G
TCIAB1185-5G
N-Boc-glycine (boc-gly-OH) ≥98.0% (by titrimetric analysis)
N-Boc-glycine (boc-gly-OH)
Formula:
C₇H₁₃NO₄ |
MDL Number:
MFCD00002690 CAS Number: 4530-20-5 EINECS: 224-864-5 |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 87.0 to 90.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...