(±)-3-Amino-3-phenylpropionic acid ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI
Formula:
C₉H₁₁NO₂ Punto di fusione: 223°C |
MDL Number:
MFCD00008064 CAS Number: 614-19-7 EINECS: 210-371-2 |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...