Stampa…

N,N-Dimethylcyanoacetamide ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-Cyano-N,N-dimethylacetamide

C1793-25G C1793-5G
TCIAC1793-25GEA 251 EUR
TCIAC1793-25G TCIAC1793-5G
N,N-Dimethylcyanoacetamide ≥98.0% (by GC)
N,N-Dimethylcyanoacetamide
Formula: C₅H₈N₂O
Peso molecolare: 112,13 g/mol
Punto di fusione: 66°C
MDL Number: MFCD00051623
CAS Number: 7391-40-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=23d1598830b54b0eefd321dbc3b10ba636d752d4&vid=23d1598830b54b0eefd321dbc3b10ba636d752d4&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 62.0 to 66.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR