Stampa…

(S)-(+)-Ethyl-3-hydroxybutyrate ≥96.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: (S)-Ethyl-3-hydroxybutyrate, Ethyl (S)-3-hydroxybutanoate, Ethyl (S)-(+)-3-hydroxybutyrate, (S)-Ethyl 3-hydroxybutanoate, Ethyl (S)-3-hydroxybutyrate

H0975-25G H0975-5G
TCIAH0975-25GEA 401 EUR
TCIAH0975-25G TCIAH0975-5G
(S)-(+)-Ethyl-3-hydroxybutyrate ≥96.0% (by GC)
(S)-(+)-Ethyl-3-hydroxybutyrate
Formula: C₆H₁₂O₃
Peso molecolare: 132,16 g/mol
Flash Pt: 64°C
MDL Number: MFCD00066206
CAS Number: 56816-01-4
EINECS: 260-393-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=26e1108352b069a131621b13105159c6c3bfdaa5&vid=26e1108352b069a131621b13105159c6c3bfdaa5&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 96.0 %
Specific rotation [a]20/D +16.0 to +19.0 deg (neat)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR