Denatonium benzoate
Fornitore: Thermo Fisher Scientific
|
Attenzione
|
CAS No.: 3734-33-6
Molecular Formula: C21H29N2O.C7H5O2
Formula Weight: 446.59
Physical Form: Powder
Appearance: White
Melting Point: 164-169°
Beilstein Reference: 8179408
Merck Reference: 14,2891
MDL No.: MFCD00031578
The bitterest compound known
Formula:
C₂₈H₃₄N₂O₃ Punto di fusione: 164…169 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00031578 CAS Number: 3734-33-6 EINECS: 223-095-2 Merck Index: 14,2891 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...