Stampa…

2-(4-Hydroxyphenylazo)benzoic acid ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-Hydroxyazobenzene-2'-carboxylic acid, 2-(4-Hydroxyphenylazo)benzoic acid, 4'-Hydroxyazobenzene-2-carboxylic acid, 2-((4-Hydroxyphenyl)diazenyl)benzoic acid, HABA

H0586-25G H0586-5G
TCIAH0586-25GEA 140 EUR
TCIAH0586-25G TCIAH0586-5G
2-(4-Hydroxyphenylazo)benzoic acid ≥98.0% (by HPLC, titration analysis)
2-(4-Hydroxyphenylazo)benzoic acid
Formula: C₁₃H₁₀N₂O₃
Peso molecolare: 242,23 g/mol
MDL Number: MFCD00002428
CAS Number: 1634-82-8
EINECS: 216-655-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8c06f8e65bc0eea5d1d4834a22da69d4dcd072da&vid=8c06f8e65bc0eea5d1d4834a22da69d4dcd072da&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR