Stampa…

(2-Methoxyphenyl)acetic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

M0533-25G M0533-5G
TCIAM0533-25GEA 41 EUR
TCIAM0533-25G TCIAM0533-5G
(2-Methoxyphenyl)acetic acid ≥98.0% (by GC, titration analysis)
(2-Methoxyphenyl)acetic acid
Formula: (C₆H₄OCH₃)CH₂COOH
Punto di ebollizione: 332 °C (1013 hPa)
Punto di fusione: 120…123 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00004321
CAS Number: 93-25-4
EINECS: 202-231-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=24efc8acfe3e636d0c87732cc55e1d15df4c9ca5&vid=24efc8acfe3e636d0c87732cc55e1d15df4c9ca5&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 123.0 to 125.0 deg-C
Solubility in Methanol very faint turbidity

Specifiche

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR