Stampa…

3-(1-Piperidinylmethyl)phenol ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3-(Piperidin-1-ylmethyl)phenol

H1183-25G H1183-5G
TCIAH1183-25GEA 357 EUR
TCIAH1183-25G TCIAH1183-5G
3-(1-Piperidinylmethyl)phenol ≥98.0% (by GC)
3-(1-Piperidinylmethyl)phenol
Formula: C₁₂H₁₇NO
Peso molecolare: 191,27 g/mol
MDL Number: MFCD00800250
CAS Number: 73279-04-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7b361d8a3b989d674b943ecb196084f9fea58acc&vid=7b361d8a3b989d674b943ecb196084f9fea58acc&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 137.0 to 141.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR