N,N-Dimethylformamide dineopentyl acetal ≥96.0% (by GC) for esterification
Fornitore: TCI
|
Attenzione
|
D1595-25ML
D1595-5ML
TCIAD1595-25MLEA
103
EUR
TCIAD1595-25ML
TCIAD1595-5ML
N,N-Dimethylformamide dineopentyl acetal ≥96.0% (by GC) for esterification
N,N-Dimethylformamide dineopentyl acetal
Formula:
C₁₃H₂₉NO₂ Punto di ebollizione: 87°C Flash Pt: 63°C |
MDL Number:
MFCD00008851 CAS Number: 4909-78-8 EINECS: 225-536-4 |
|
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...