Stampa…

1,3,5-Tris(bromomethyl)benzene ≥98.0% (by GC)

Fornitore: TCI
Pericolo

T2719-1G T2719-5G
TCIAT2719-1GEA 33 EUR
TCIAT2719-1G TCIAT2719-5G
1,3,5-Tris(bromomethyl)benzene ≥98.0% (by GC)
1,3,5-Tris(bromomethyl)benzene
Formula: C₉H₉Br₃
MDL Number: MFCD00182500
CAS Number: 18226-42-1
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=853f072c7445cf6a92405845570d2964e608d0f2&vid=853f072c7445cf6a92405845570d2964e608d0f2&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 97.0 to 101.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR