Stampa…

4-Fluoro-N-methylbenzylamine ≥97.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: N-Methyl-4-fluorobenzylamine

F0806-1G F0806-5G
TCIAF0806-1GEA 28 EUR
TCIAF0806-1G TCIAF0806-5G
4-Fluoro-N-methylbenzylamine ≥97.0% (by GC, titration analysis)
4-Fluoro-N-methylbenzylamine
Formula: C₈H₁₀FN
Peso molecolare: 139,17 g/mol
Punto di ebollizione: 75°C
Flash Pt: 71°C
MDL Number: MFCD03839848
CAS Number: 405-66-3
UN: 2735
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=414ba7f96084717e1a6a15b88c9647502c3ee249&vid=414ba7f96084717e1a6a15b88c9647502c3ee249&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR