1-(2-Bromoethoxy)-2-ethoxybenzene ≥96.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2-(2-Ethoxyphenoxy)ethyl bromide
B3191-25G
B3191-5G
TCIAB3191-25GEA
104
EUR
TCIAB3191-25G
TCIAB3191-5G
1-(2-Bromoethoxy)-2-ethoxybenzene ≥96.0% (by GC)
1-(2-Bromoethoxy)-2-ethoxybenzene
Formula:
C₁₀H₁₃BrO₂ Peso molecolare: 245,12 g/mol |
MDL Number:
MFCD02030483 CAS Number: 3259-03-8 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 96.0 % |
Melting point | 41.0 to 45.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...