3',4'-Dimethoxy-2'-hydroxyacetophenone ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Gallacetophenone 3',4'-dimethyl ether
H1205-25G
H1205-5G
TCIAH1205-25GEA
312
EUR
TCIAH1205-25G
TCIAH1205-5G
3',4'-Dimethoxy-2'-hydroxyacetophenone ≥98.0% (by GC)
3',4'-Dimethoxy-2'-hydroxyacetophenone
Formula:
C₁₀H₁₂O₄ |
MDL Number:
MFCD00016684 CAS Number: 5396-18-9 |
Risultati del test di specificazione
Purity (GC) | min. 98.0 % |
Melting point | 75.0 to 79.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...