Stampa…

3-(Benzyloxy)phenol ≥95.0% (by GC)

Fornitore: TCI
Attenzione

B2070-25G B2070-5G
TCIAB2070-25GEA 398 EUR
TCIAB2070-25G TCIAB2070-5G
3-(Benzyloxy)phenol ≥95.0% (by GC)
3-(Benzyloxy)phenol
Formula: C₁₃H₁₂O₂
Punto di ebollizione: 200°C
MDL Number: MFCD00134682
CAS Number: 3769-41-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba1930815c9aa496106eade3b02b776f11358163&vid=ba1930815c9aa496106eade3b02b776f11358163&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Melting point 50.0 to 54.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR