Stampa…

Trichromium dicarbide ≥99.5% (metals basis)

Fornitore: Thermo Fisher Scientific
Attenzione

11547.18 11547.30
11547.18EA 59.5 EUR
11547.18 11547.30
Trichromium dicarbide ≥99.5% (metals basis)
Trichromium dicarbide

Purity: 99.5% CAS No.: 12012-35-0 Molecular Formula: Cr3C2 Formula Weight: 180.02 Physical Form: -325 Mesh Powder Odour: Odourless UN No.: 3178 MDL No.: MFCD00016041

Formula: C₂Cr₃
Punto di fusione: 1890 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00016041
CAS Number: 12012-35-0
EINECS: 234-576-1
UN: 3178
ADR: 4.1,III

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR