Stampa…

4'-Chloro-2,2,2-trifluoroacetophenone ≥97.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 1-(4-Chlorophenyl)-2,2,2-trifluoroethanone

C1738-1G C1738-5G
TCIAC1738-1GEA 31 EUR
TCIAC1738-1G TCIAC1738-5G
4'-Chloro-2,2,2-trifluoroacetophenone ≥97.0% (by GC)
4'-Chloro-2,2,2-trifluoroacetophenone
Formula: C₈H₄ClF₃O
Peso molecolare: 208,5700 g/mol
MDL Number: MFCD00018686
CAS Number: 321-37-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7db49b3de521183144f587c95a5f7be963c2666e&vid=7db49b3de521183144f587c95a5f7be963c2666e&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to lump
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR