2,4,6-Triisopropylphenylboronic acid ≥98%
Fornitore: Thermo Scientific
|
Attenzione
|
Synonyms:
2,4,6-Triisopropylbenzeneboronic acid, (2,4,6-Triisopropylphenyl)boronic acid
B22891.03
B22891.06
B22891.14
B22891.03EA
71.4
EUR
B22891.03
B22891.06
B22891.14
2,4,6-Triisopropylphenylboronic acid ≥98%
2,4,6-Triisopropylphenylboronic acid
Purity: 98%
CAS No.: 154549-38-9
Molecular Formula: C15H25BO2
Formula Weight: 248.16
Melting Point: ca 165° dec.
MDL No.: MFCD02683099
Formula:
C₁₅H₂₅BO₂ Peso molecolare: 248,16 g/mol Punto di fusione: ∼ 165 °C (dec.) Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD02683099 CAS Number: 154549-38-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...