Stampa…

2-Chlorothioxanthone ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-Chlorothioxanthen-9-one, 2-Chloro-9H-thioxanthen-9-one

C0292-25G C0292-5G
TCIAC0292-25GEA 94 EUR
TCIAC0292-25G TCIAC0292-5G
2-Chlorothioxanthone ≥98.0% (by GC)
2-Chlorothioxanthone
Formula: C₁₃H₇ClOS
Peso molecolare: 246,72 g/mol
MDL Number: MFCD00005067
CAS Number: 86-39-5
EINECS: 201-667-2
UN: 1325
ADR: 4.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57e7ed4c3cdb92180056125497bc8141a7b181e8&vid=57e7ed4c3cdb92180056125497bc8141a7b181e8&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 150.0 to 153.0 deg-C
Solubility in hot Toluene very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR