Stampa…

Isoquinolin-N-oxide

Fornitore: TCI EUROPE

Synonyms: Isoquinoline N-Oxide

I0540-25G I0540-5G
TCIAI0540-25GEA 639 EUR
TCIAI0540-25G TCIAI0540-5G
Isoquinolin-N-oxide
Isoquinolin-N-oxide
Formula: C₉H₇NO
Peso molecolare: 145,16 g/mol
MDL Number: MFCD00006903
CAS Number: 1532-72-5
EINECS: 216-242-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=49f5eaa117dd7162a31b640d55ce76ef8c4f7b6f&vid=49f5eaa117dd7162a31b640d55ce76ef8c4f7b6f&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 103.0 to 105.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR