4-Biphenylboronic acid (contains varying amounts of Anhydride)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
[1,1'-Biphenyl]-4-ylboronic acid
B2294-25G
B2294-5G
TCIAB2294-25GEA
62
EUR
TCIAB2294-25G
TCIAB2294-5G
4-Biphenylboronic acid (contains varying amounts of Anhydride)
4-Biphenylboronic acid
Formula:
C₁₂H₁₁BO₂ Peso molecolare: 198,03 g/mol Punto di fusione: 266°C |
MDL Number:
MFCD00093311 CAS Number: 5122-94-1 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (Neutralization titration) | 97.0 to 110.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...