Stampa…

3',5'-Diacetoxyacetophenone ≥98.0% (by GC)

Fornitore: TCI

Synonyms: 5-Acetyl-1,3-phenylene diacetate

D1978-10G D1978-25G D1978-5G
TCIAD1978-10GEA 50 EUR
TCIAD1978-10G TCIAD1978-25G TCIAD1978-5G
3',5'-Diacetoxyacetophenone ≥98.0% (by GC)
3',5'-Diacetoxyacetophenone
Formula: C₁₂H₁₂O₅
Peso molecolare: 236.2207 g/mol
MDL Number: MFCD00008702
CAS Number: 35086-59-0
EINECS: 252-354-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=497c4b203ae335ab300b2bef77636aaaf9ccd3bc&vid=497c4b203ae335ab300b2bef77636aaaf9ccd3bc&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 91.0 to 95.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR