N-Fluorobenzenesulphonimide ≥97%
Fornitore: Thermo Scientific
|
Attenzione
|
L13955.03
L13955.06
L13955.14
L13955.03EA
21.1
EUR
L13955.03
L13955.06
L13955.14
N-Fluorobenzenesulphonimide ≥97%
N-Fluorobenzenesulphonimide
Purity: 97%
CAS No.: 133745-75-2
Molecular Formula: C12H10FNO4S2
Formula Weight: 315.34
Melting Point: 114-118°
Beilstein Reference: 5348902
Formula:
C₁₂H₁₀FNO₄S₂ Peso molecolare: 315,34 g/mol Punto di fusione: 114…118 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00144885 CAS Number: 133745-75-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...