2-Nitrophenyl phenyl sulphide ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
N0744-25G
N0744-5G
TCIAN0744-25GEA
185
EUR
TCIAN0744-25G
TCIAN0744-5G
2-Nitrophenyl phenyl sulphide ≥98.0% (by GC)
2-Nitrophenyl phenyl sulphide
Formula:
C₁₂H₉NO₂S |
CAS Number:
4171-83-9 |
Risultati del test di specificazione
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 80.0 to 84.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...