Stampa…

4-Carboxy-3-fluorophenylboronic acid ≥98%

Fornitore: Thermo Scientific
Attenzione

Synonyms: 4-Borono-2-fluorobenzoic acid, 4-Carboxy-3-fluorobenzeneboronic acid

H53285.03 H53285.06
H53285.03EA 53.5 EUR
H53285.03 H53285.06
4-Carboxy-3-fluorophenylboronic acid ≥98%
4-Carboxy-3-fluorophenylboronic acid

Purity: 98% CAS No.: 120153-08-4 Molecular Formula: C7H6BFO4 Formula Weight: 183.93 Melting Point: 236-240° MDL No.: MFCD01114671

Formula: C₇H₆BFO₄
Peso molecolare: 183,93 g/mol
Punto di fusione: 236…240 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD01114671
CAS Number: 120153-08-4

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR