Stampa…

4-Fluoro-3-(methylcarbamoyl)phenylboronic acid 96%

Fornitore: Thermo Scientific
Attenzione

Synonyms: (4-Fluoro-3-(methylcarbamoyl)phenyl)boronic acid, 4-Fluoro-3-(methylcarbamoyl)benzeneboronic acid

H53101.MD H53101.03
H53101.MDEA 160 EUR
H53101.MD H53101.03
4-Fluoro-3-(methylcarbamoyl)phenylboronic acid 96%
4-Fluoro-3-(methylcarbamoyl)phenylboronic acid

Purity: 96% CAS No.: 874219-19-9 Molecular Formula: C8H9BFNO3 Formula Weight: 196.97 Melting Point: 226-228° MDL No.: MFCD08235038

Formula: C₈H₉BFNO₃
Peso molecolare: 196,97 g/mol
Punto di fusione: 226…228 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD08235038
CAS Number: 874219-19-9

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR