Stampa…

2-Amino-4-chloro-6-nitrophenol ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

A2244-100G A2244-25G
TCIAA2244-100GEA 139 EUR
TCIAA2244-100G TCIAA2244-25G
2-Amino-4-chloro-6-nitrophenol ≥98.0% (by GC, titration analysis)
2-Amino-4-chloro-6-nitrophenol
Formula: C₆H₅ClN₂O₃
Peso molecolare: 188,57 g/mol
Punto di fusione: 152°C
MDL Number: MFCD00035925
CAS Number: 6358-08-3
EINECS: 228-761-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=22bad03e64cb6cbe0b92c551033ad1e2a60a657f&vid=22bad03e64cb6cbe0b92c551033ad1e2a60a657f&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Amber to Dark green powder to crystaline
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 145.0 to 149.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR