2-(Ethoxycarbonyl)phenylboronic acid (contains varying amounts of Anhydride)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2-(Ethoxycarbonyl)benzeneboronic acid, (2-(Ethoxycarbonyl)phenyl)boronic acid
E0877-1G
E0877-5G
TCIAE0877-1GEA
20
EUR
TCIAE0877-1G
TCIAE0877-5G
2-(Ethoxycarbonyl)phenylboronic acid (contains varying amounts of Anhydride)
2-(Ethoxycarbonyl)phenylboronic acid
Formula:
C₉H₁₁BO₄ Peso molecolare: 193,99 g/mol Punto di fusione: 138°C |
MDL Number:
MFCD02179453 CAS Number: 380430-53-5 |
Risultati del test di specificazione
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (Neutralization titration) | 95.0 to 110.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...