Stampa…

(S)-(-)-N-Benzyl-α-methylbenzylamine ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: S-(-)-N-Benzyl-1-phenylethylamine, (S)-(-)-N-Benzyl-1-phenylethylamine

B1529-25ML B1529-5ML
TCIAB1529-25MLEA 57 EUR
TCIAB1529-25ML TCIAB1529-5ML
(S)-(-)-N-Benzyl-α-methylbenzylamine ≥98.0% (by GC, titration analysis)
(S)-(-)-N-Benzyl-α-methylbenzylamine
Formula: C₁₅H₁₇N
Peso molecolare: 211,31 g/mol
MDL Number: MFCD00066325
CAS Number: 17480-69-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=642a4882ff56340a6ff71f18474493ed3a6d0a73&vid=642a4882ff56340a6ff71f18474493ed3a6d0a73&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D -38.0 to -41.0 deg (neat)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR