Stampa…

DL-Methionine sulphone ≥99.0% (by titrimetric analysis)

Fornitore: TCI EUROPE

M0100-1G M0100-5G
TCIAM0100-1GEA 30 EUR
TCIAM0100-1G TCIAM0100-5G
DL-Methionine sulphone ≥99.0% (by titrimetric analysis)
DL-Methionine sulphone
Formula: C₅H₁₁NO₄S
Peso molecolare: 181,21 g/mol
MDL Number: MFCD00025079
CAS Number: 820-10-0
EINECS: 212-466-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=73febbdb144c2198e00f8cde83c5fba4f872089f&vid=73febbdb144c2198e00f8cde83c5fba4f872089f&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 99.0 %
Solubility in hot Water very faint turbidit

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR