Stampa…

Benzothiazol-2-ol ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 2-Hydroxybenzothiazole

B2344-25G B2344-5G
TCIAB2344-25GEA 85 EUR
TCIAB2344-25G TCIAB2344-5G
Benzothiazol-2-ol ≥98.0% (by GC)
Benzothiazol-2-ol
Formula: C₇H₅NOS
Peso molecolare: 151,18 g/mol
MDL Number: MFCD00022868
CAS Number: 934-34-9
EINECS: 213-281-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d9e3e16e687662be841c8a8ed84be822efee576&vid=67f0eaca722bc075054e61d9f31518fe7f761d4f&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 136.0 to 141.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR