Stampa…

Tetrathiafulvalene ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 2,2'-Bi(1,3-dithiolylidene), TTF

T0980-1G T0980-25G T0980-5G
TCIAT0980-1GEA 90 EUR
TCIAT0980-1G TCIAT0980-25G TCIAT0980-5G
Tetrathiafulvalene ≥98.0% (by GC)
Tetrathiafulvalene
Formula: C₆H₄S₄
Peso molecolare: 204,356 g/mol
MDL Number: MFCD00005492
CAS Number: 31366-25-3
EINECS: 250-593-7
Merck Index: 14,09242
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=873f56551091dec6425e81c90c521c2dfe179ba4&vid=873f56551091dec6425e81c90c521c2dfe179ba4&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Orange to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 118.0 to 123.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR