Stampa…
Outdoor jacket, Oban S523
Fornitore: Portwest
S523NAR1XL S523NAR2XL S523NAR3XL S523NARL S523NARM S523NARS
113-7523EA 62.6 EUR
113-7523 113-7524 113-7525 113-7526 113-7527 113-7528
Outdoor jacket, Oban S523
Abbigliamento da lavoro
This waterproof polyester jacket with PVC coating is ideal for outdoor activities. It features a polyester fleece lining, and an additional nylon lining in the upper body and the sleeves.
  • Two hip pockets with zips, mobile phone pocket
  • Taped waterproof seams, two-way zip front with storm flap
  • Drawstring hood and waist
  • Adjustable Velcro® cuffs
Weight: 160 g/m²; fleece 280 g/m²
EN 343 (3-1)
Certificazioni: EN 343 (3-1)